PRI424050386 Princeton BioMolecular Research, Inc.

7-Chloro-5-cyclobutyl-1,3-dimethyl-1H-pyrazolo[4,3-d]pyrimidine

  • MDL number MFCD14704955
  • Cas Number : 1239783-12-0
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart