PRI424050501 Princeton BioMolecular Research, Inc.

5-Ethoxy-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09835050
  • Cas Number : 1196157-35-3
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-ethoxy-1-methyl-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C7H10N2O3/c1-3-12-6-4-5(7(10)11)8-9(6)2/h4H,3H2,1-2H3,(H,10,11)
Inchi Key PDWVNFTVXUYADL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart