PRI424050502 Princeton BioMolecular Research, Inc.

5-Isopropoxy-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD16547218
  • Cas Number : 1239783-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart