PRI424050673 Princeton BioMolecular Research, Inc.

2-Chloro-6-methyl-pyridine-3,5-dicarboxylic acid 3-ethyl ester 5-methyl ester

  • MDL number MFCD16630783
  • Cas Number : 1239731-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart