PRI424050683 Princeton BioMolecular Research, Inc.

2-(3-Ethyl-[1,2,4]oxadiazol-5-yl)-piperidine

  • MDL number MFCD11116244
  • Cas Number : 1036454-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart