PRI424050689 Princeton BioMolecular Research, Inc.

3-Formyl-1-methyl-1H-indole-4-carboxylic acid methyl ester

  • MDL number MFCD09864027
  • Cas Number : 65923-20-8
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart