PRI424050691 Princeton BioMolecular Research, Inc.

6-Methyl-1,2,3,9-tetrahydro-carbazol-4-one

  • MDL number MFCD11168367
  • Cas Number : 51626-88-1
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
Inchi Code 1S/C13H13NO/c1-8-5-6-10-9(7-8)13-11(14-10)3-2-4-12(13)15/h5-7,14H,2-4H2,1H3
Inchi Key YCYFUAOBKJMHPH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart