PRI424050702 Princeton BioMolecular Research, Inc.

4-Methoxy-6-methyl-5,6,7,8-tetrahydro-[1,3]dioxolo[4,5-g]isoquinoline

  • MDL number MFCD00868501
  • Cas Number : 550-10-7
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart