PRI424050721 Princeton BioMolecular Research, Inc.

1-Ethyl-3,3-dimethyl-5-nitro-1,3-dihydro-indol-2-one

  • MDL number MFCD16994188
  • Cas Number : 135437-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart