PRI424050722 Princeton BioMolecular Research, Inc.

1,3,3-Trimethyl-5-nitro-1,3-dihydro-indol-2-one

  • MDL number MFCD03147762
  • Cas Number : 120791-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart