PRI424050749 Princeton BioMolecular Research, Inc.

2-[4-(4-Chloro-phenyl)-thiazol-2-yl]-ethylamine

  • MDL number MFCD05221116
  • Cas Number : 641993-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart