PRI424050765 Princeton BioMolecular Research, Inc.

2-(3,3,5-trimethyl-2-oxo-indolin-1-yl)acetic acid

  • MDL number MFCD17265877
  • Cas Number : 1248550-14-2
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,3,5-trimethyl-2-oxo-2,3-dihydro-1H-indol-1-yl)acetic acid
Inchi Code 1S/C13H15NO3/c1-8-4-5-10-9(6-8)13(2,3)12(17)14(10)7-11(15)16/h4-6H,7H2,1-3H3,(H,15,16)
Inchi Key RUELKNITWFAFGV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart