PRI424050770 Princeton BioMolecular Research, Inc.

1,3,3,7-tetramethylindolin-2-one

  • MDL number MFCD17265881
  • Cas Number : 344287-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart