PRI424050809 Princeton BioMolecular Research, Inc.

2-[3-(4-Chloro-phenyl)-[1,2,4]oxadiazol-5-yl]-ethylamine

  • MDL number MFCD06738202
  • Cas Number : 885953-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart