PRI424050814 Princeton BioMolecular Research, Inc.

1-(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-1H-[1,2,3]triazole-4,5-dicarboxylic acid

  • MDL number MFCD18088338
  • Molecular Weight : 291.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart