PRI424050926 Princeton BioMolecular Research, Inc.

6-Methoxy-2H-chromene

  • MDL number MFCD01662550
  • Cas Number : 18385-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart