PRI424050953 Princeton BioMolecular Research, Inc.

2-[4-(Propane-2-sulfonyl)-piperazin-1-yl]-ethylamine

  • MDL number MFCD12798991
  • Molecular Weight : 235.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[4-(isopropylsulfonyl)-1-piperazinyl]ethanamine
Inchi Code 1S/C9H21N3O2S/c1-9(2)15(13,14)12-7-5-11(4-3-10)6-8-12/h9H,3-8,10H2,1-2H3
Inchi Key HTNRLEHTEKSUPH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart