PRI424050962 Princeton BioMolecular Research, Inc.

4-Amino-3-(4-fluoro-phenyl)-butyric acid methyl ester

  • MDL number MFCD18324297
  • Molecular Weight : 211.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-amino-3-(4-fluorophenyl)butanoate
Inchi Code 1S/C11H14FNO2/c1-15-11(14)6-9(7-13)8-2-4-10(12)5-3-8/h2-5,9H,6-7,13H2,1H3
Inchi Key VNRMYROTJKSLET-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart