PRI424051229 Princeton BioMolecular Research, Inc.

methyl 6-methyl-2-pyrrolidin-1-yl-pyrimidine-4-carboxylate

  • MDL number MFCD18325124
  • Cas Number : 1263215-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart