PRI424051231 Princeton BioMolecular Research, Inc.

methyl 2-(dimethylamino)-6-methyl-pyrimidine-4-carboxylate

  • MDL number MFCD18325126
  • Cas Number : 1263209-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart