PRI424051322 Princeton BioMolecular Research, Inc.

2-(4-Trifluoromethoxy-phenyl)-cyclopropanecarboxylic acid

  • MDL number MFCD12068247
  • Cas Number : 1157562-31-6
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart