PRI424051339 Princeton BioMolecular Research, Inc.

(3-Chloro-phenyl)-propynoic acid

  • MDL number MFCD14707279
  • Cas Number : 7396-28-3
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-chlorophenyl)-2-propynoic acid
Inchi Code 1S/C9H5ClO2/c10-8-3-1-2-7(6-8)4-5-9(11)12/h1-3,6H,(H,11,12)
Inchi Key FRTZPNZKFMZQDR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart