PRI424051359 Princeton BioMolecular Research, Inc.

(5-Hydroxy-isoxazol-3-yl)-acetic acid

  • MDL number MFCD09751674
  • Cas Number : 178168-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart