PRI424051463 Princeton BioMolecular Research, Inc.

3-Amino-N-isopropyl-propionamide

  • MDL number MFCD09036524
  • Cas Number : 937690-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart