PRI424051473 Princeton BioMolecular Research, Inc.

3-Amino-4-(4-chloro-phenyl)-1-methyl-azetidin-2-one

  • MDL number MFCD18838160
  • Cas Number : 1291488-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart