PRI424051819 Princeton BioMolecular Research, Inc.

2-[4-(2-methoxyethylsulfonyl)piperazin-1-yl]ethanamine

  • MDL number MFCD16059736
  • Molecular Weight : 251.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{4-[(2-methoxyethyl)sulfonyl]-1-piperazinyl}ethanamine
Inchi Code 1S/C9H21N3O3S/c1-15-8-9-16(13,14)12-6-4-11(3-2-10)5-7-12/h2-10H2,1H3
Inchi Key FJYIHFIHMYGPQU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart