PRI424051825 Princeton BioMolecular Research, Inc.

6-Bromo-2-(2-methoxy-ethyl)-2,3-dihydro-isoindol-1-one

  • MDL number MFCD20441054
  • Cas Number : 1344692-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart