PRI424052643 Princeton BioMolecular Research, Inc.

3-{[3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-methoxybenzaldehyde

  • MDL number MFCD03419736
  • Cas Number : 1001518-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart