PRI424052844 Princeton BioMolecular Research, Inc.

ethyl 5-(chlorocarbonyl)-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD03421623
  • Cas Number : 1197231-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart