PRI424053696 Princeton BioMolecular Research, Inc.

1-(4-chloro-1-ethyl-1H-pyrazol-3-yl)ethanone

  • MDL number MFCD04968844
  • Cas Number : 1006434-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart