PRI424053908 Princeton BioMolecular Research, Inc.

3-{[4-amino-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}benzoic acid

  • MDL number MFCD04969613
  • Cas Number : 1006471-27-7
  • Molecular Weight : 285.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-{[4-amino-3-(trifluoromethyl)-1H-pyrazol-1-yl]methyl}benzoic acid
Inchi Code 1S/C12H10F3N3O2/c13-12(14,15)10-9(16)6-18(17-10)5-7-2-1-3-8(4-7)11(19)20/h1-4,6H,5,16H2,(H,19,20)
Inchi Key IFJBIRQVTXAGAU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart