PRI424054989 Princeton BioMolecular Research, Inc.

5-(chloromethyl)-3-(1-methyl-1H-pyrazol-4-yl)isoxazole

  • MDL number MFCD06805624
  • Cas Number : 1006483-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart