PRI424055087 Princeton BioMolecular Research, Inc.

1-[(1-ethyl-3-methyl-1H-pyrazol-4-yl)sulfonyl]piperidin-4-amine

  • MDL number MFCD06805733
  • Cas Number : 1018996-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart