PRI424055409 Princeton BioMolecular Research, Inc.

6-chloro-1-(4-methylphenyl)-2,3,4,9-tetrahydro-1H-beta-carboline

  • MDL number MFCD05190845
  • Cas Number : 1431964-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart