PRI424055457 Princeton BioMolecular Research, Inc.

5-(6-cyclopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-4-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08553253
  • Cas Number : 937598-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart