PRI424055727 Princeton BioMolecular Research, Inc.

ethyl 3-(3-ethyl[1,2,4]triazolo[3,4-b][1,3,4]thiadiazol-6-yl)-1-methyl-1H-pyrazole-5-carboxylate

  • MDL number MFCD08557673
  • Cas Number : 1006326-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart