PRI424055862 Princeton BioMolecular Research, Inc.

1-(2,2-difluoroethyl)piperidine-4-carboxylic acid

  • MDL number MFCD08696502
  • Cas Number : 937602-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart