PRI424055917 Princeton BioMolecular Research, Inc.

N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]-N-(2-methoxybenzyl)amine

  • MDL number MFCD08696825
  • Cas Number : 1006473-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart