PRI424055918 Princeton BioMolecular Research, Inc.

N-(2,3-dimethoxybenzyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]amine

  • MDL number MFCD08696826
  • Cas Number : 1006437-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart