PRI424055974 Princeton BioMolecular Research, Inc.

N,N-bis[(1-methyl-1H-pyrazol-4-yl)methyl]amine

  • MDL number MFCD08558997
  • Cas Number : 1006446-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart