PRI424056052 Princeton BioMolecular Research, Inc.

4-[(1-ethyl-3-methyl-1H-pyrazol-4-yl)methoxy]-3-fluoroaniline

  • MDL number MFCD08699611
  • Cas Number : 1006961-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart