PRI424056068 Princeton BioMolecular Research, Inc.

1-[4-(2-phenyl-1H-imidazol-1-yl)phenyl]methanamine

  • MDL number MFCD08699671
  • Cas Number : 937598-81-7
  • Molecular Weight : 249.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart