PRI424056084 Princeton BioMolecular Research, Inc.

2-[(1-methyl-1H-pyrazol-4-yl)methoxy]pyridin-3-amine

  • MDL number MFCD08700199
  • Cas Number : 1006464-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart