PRI424056085 Princeton BioMolecular Research, Inc.

6-[(1-methyl-1H-pyrazol-4-yl)methoxy]pyridin-3-amine

  • MDL number MFCD08700245
  • Cas Number : 1432027-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart