PRI424056090 Princeton BioMolecular Research, Inc.

5-chloro-2-[(1-methyl-1H-pyrazol-4-yl)methoxy]aniline

  • MDL number MFCD08696915
  • Cas Number : 1006962-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart