PRI424056226 Princeton BioMolecular Research, Inc.

4,4-difluoro-1-(1-methyl-1H-pyrazol-3-yl)butane-1,3-dione

  • MDL number MFCD08559027
  • Cas Number : 1006333-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart