PRI424056346 Princeton BioMolecular Research, Inc.

2-methyl-4H-furo[3,4-c]pyrazole-4,6(2H)-dione

  • MDL number MFCD09473432
  • Cas Number : 1011396-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart