PRI424056391 Princeton BioMolecular Research, Inc.

methyl 1-(2,2,3,3-tetrafluoropropyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD09473425
  • Cas Number : 1172752-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart