PRI424056563 Princeton BioMolecular Research, Inc.

7-(3-aminophenyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD09701563
  • Cas Number : 1018052-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart