PRI424056718 Princeton BioMolecular Research, Inc.

5-(chloromethyl)-1-ethyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD09701717
  • Cas Number : 1171086-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart